Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(n-propylcyclopentadienyl)magnesium, 98+%

Chemical Identifiers

Chemical Name or Material Bis(n-propylcyclopentadienyl)magnesium
CAS 114504-74-4
Formula Weight 238.66
MDL Number MFCD00151384
UN Number UN3391
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3924003

 
alfa aesar™
3924003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3924006

 
alfa aesar™
3924006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39240MD

 
alfa aesar™
39240MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 238.66
Packaging Ampule
Assay ≥98%
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC
Chemical Name or Material Bis(n-propylcyclopentadienyl)magnesium
Physical Form Crystalline
MDL Number MFCD00151384
CAS 114504-74-4
UN Number UN3391
Quantity 1g
Health Hazard 1 Danger
Sensitivity Air & Moisture Sensitive
Health Hazard 2 GHS H Statement
H250-H252
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Health Hazard 3 GHS P Statement
P210-P222-P280-P370+P378a-P405-P422a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store locked up.
Store contents under inert gas.
Density 0.94g/mL
Safety and Handling

Danger

GHS P Statement
P210-P222-P280-P370+P378a-P405-P422a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store locked up.
Store contents under inert gas.

GHS H Statement
H250-H252
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.