Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(n-butylcyclopentadienyl)zirconium(IV) dichloride, 98+%

$119.30 - $455.62

Chemical Identifiers

Chemical Name or Material Bis(n-butylcyclopentadienyl)zirconium(IV) dichloride
CAS 73364-10-0
Molecular Formula C18H26Cl2Zr
Formula Weight 404.53
MDL Number MFCD00672105
Synonym 1,1'-Di-n-butylzirconocene dichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2757603

 
alfa aesar™
H2757603
1g Each for $119.30

AAH2757606

 
alfa aesar™
H2757606
5g Each for $455.62
Description & Specifications

Specifications

Formula Weight 404.53
Assay ≥98%
Chemical Name or Material Bis(n-butylcyclopentadienyl)zirconium(IV) dichloride
TSCA TSCA
Molecular Formula C18H26Cl2Zr
MDL Number MFCD00672105
CAS 73364-10-0
Synonym 1,1'-Di-n-butylzirconocene dichloride
Melting Point 97° to 99°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.