Please login to your online account to display your discounted pricing

Bis(MSB), Scintillation Grade 99.5%, ACROS Organics™

$550.10 - $550.10

Chemical Identifiers

Chemical Name or Material Bis(MSB)
Name Note 99.5%
CAS 13280-61-0
CAS Min % 99.4
CAS Max % 100.0
Assay ≥99.4%
Assay Percent Range 99.50%
Molecular Formula C24H22
Formula Weight 310.44
MDL Number MFCD00008529
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC402601000

  SDS
thermo fisher scientific
402601000
Each for $550.10
Description & Specifications

Specifications

Assay ≥99.4%
Physical Form Crystalline Powder
Formula Weight 310.44
Quantity 100g
Color Light green to yellow-green
TSCA TSCA
EINECS Number 236-285-5
Melting Point 179° to 181°C
Chemical Name or Material Bis(MSB)
Name Note 99.5%
Assay Percent Range 99.50%
MDL Number MFCD00008529
Synonym 1, 4-Bis(2-methylstyryl)benzene
CAS Min % 99.4
CAS Max % 100.0
Infrared Spectrum Authentic
Lambda Max 415nm (absolute fluorescence emission in toluene)
Molecular Formula C24H22
CAS 13280-61-0
Suitability (as a reagent in liquid scintillation counting) Passes Test

for fluorescence/luminescence spectroscopy