Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(methylthiomethyl) sulfide, 97%

$156.08 - $156.08

Chemical Identifiers

Chemical Name or Material Bis(methylthiomethyl) sulfide
CAS 6540-86-9
Molecular Formula C4H10S3
Linear Formula (CH3SCH2)2S
Formula Weight 154.30g/mol
MDL Number MFCD08436096
Synonym Thiobis(methylthio)methane; 2,4,6-Trithiaheptane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2643506

 
alfa aesar™
H26435-06
5g Each for $156.08
Description & Specifications

Specifications

Quantity 5g
Odor Stench
Chemical Name or Material Bis(methylthiomethyl) sulfide
CAS 6540-86-9
Assay 97%
Molecular Formula C4H10S3
Linear Formula (CH3SCH2)2S
Formula Weight 154.30g/mol
MDL Number MFCD08436096
Synonym Thiobis(methylthio)methane; 2,4,6-Trithiaheptane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.