Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(methylcyclopentadienyl)zirconium dichloride, 98%

Chemical Identifiers

Chemical Name or Material Bis(methylcyclopentadienyl)zirconium dichloride
CAS 12109-71-6
Molecular Formula C12H14Cl2Zr
Formula Weight 320.37
MDL Number MFCD00672106
UN Number UN3261
Synonym 1,1'-Dimethylzirconocene dichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2724103

 
alfa aesar™
H2724103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2724106

 
alfa aesar™
H2724106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 320.37
Chemical Name or Material Bis(methylcyclopentadienyl)zirconium dichloride
TSCA TSCA
Assay 98%
Molecular Formula C12H14Cl2Zr
MDL Number MFCD00672106
CAS 12109-71-6
UN Number UN3261
Synonym 1,1'-Dimethylzirconocene dichloride
Melting Point 180° to 184°C
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H260-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H260-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.