Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(methylcyclopentadienyl)nickel(II), 97%

$121.15 - $121.15

Chemical Identifiers

Chemical Name or Material Bis(methylcyclopentadienyl)nickel(II)
CAS 1293-95-4
Molecular Formula C12H14Ni
Formula Weight 216.93
MDL Number MFCD01863654
Synonym 1,1'-Dimethylnickelocene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2746403

 
alfa aesar™
H2746403
1g Each for $121.15
Description & Specifications

Specifications

Formula Weight 216.93
Chemical Name or Material Bis(methylcyclopentadienyl)nickel(II)
Assay 97%
Molecular Formula C12H14Ni
MDL Number MFCD01863654
CAS 1293-95-4
Synonym 1,1'-Dimethylnickelocene
Melting Point 34° to 36°C
Boiling Point 85° to 90°C (1mmHg)
Quantity 1g
Flash Point 66°C (151°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H351-H332-H317
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Harmful if inhaled.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H351-H332-H317
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Harmful if inhaled.
May cause an allergic skin reaction.

2° to 8°C