Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(mercaptocyclohexane)titanium tetrachloride, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Bis(mercaptocyclohexane)titanium tetrachloride
CAS 149391-23-1
Formula Weight 422.1
MDL Number MFCD00171702
UN Number UN3261
Synonym Tetrachlorobis(cyclohexylmercapto)titanium(IV)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3954003

  SDS
alfa aesar™
3954003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3954006

  SDS
alfa aesar™
3954006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 422.1
Chemical Name or Material Bis(mercaptocyclohexane)titanium tetrachloride
Physical Form Crystalline
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00171702
CAS 149391-23-1
UN Number UN3261
Synonym Tetrachlorobis(cyclohexylmercapto)titanium(IV)
Color Yellow
Melting Point 76° to 80°C
Quantity 1g
Health Hazard 1 Danger
Sensitivity Moisture Sensitive
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.