Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(isopropylcyclopentadienyl)zirconium dichloride, 98%

$77.59 - $77.59

Chemical Identifiers

Chemical Name or Material Bis (isopropylcyclopentadienyl)zirconium dichloride
CAS 58628-40-3
Molecular Formula C16H22Cl2Zr
Formula Weight 376.48
MDL Number MFCD00269855
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4323202

  SDS
alfa aesar™
4323202
0.5g Each for $77.59
Description & Specifications

Specifications

Formula Weight 376.48
Chemical Name or Material Bis (isopropylcyclopentadienyl)zirconium dichloride
Assay 98%
Molecular Formula C16H22Cl2Zr
Physical Form Powder
Melting Point 134° to 137°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Moisture Sensitive
Quantity 0.5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 58628-40-3
Health Hazard 1 Warning
MDL Number MFCD00269855
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.