Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(isopropylcyclopentadienyl)tungsten dihydride, 98+%

MOCVD precursor

Chemical Identifiers

Chemical Name or Material Bis (isopropylcyclopentadienyl)tungsten dihydride
CAS 64561-25-7
Formula Weight 400.22
MDL Number MFCD04126431
UN Number UN3398
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4267202

  SDS
alfa aesar™
4267202
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4267204

  SDS
alfa aesar™
4267204
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Bis (isopropylcyclopentadienyl)tungsten dihydride
CAS 64561-25-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H261-H315-H319-H335
Flammable liquid and vapor.
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
DOT Information Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE
Packaging Ampule
Physical Form Liquid
Assay ≥98%
Formula Weight 400.22
MDL Number MFCD04126431
UN Number UN3398
Color Orange
Boiling Point 124°C (0.1mmHg)
Quantity 0.5g
Sensitivity Air & Moisture Sensitive

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H261-H315-H319-H335
Flammable liquid and vapor.
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Ampouled under argon