Please login to your online account to display your discounted pricing

Bis(isopropylcyclopentadienyl)hafnium dichloride, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Bis(isopropylcyclopentadienyl)hafnium dichloride
CAS 66349-80-2
Molecular Formula C16H22Cl2Hf
Formula Weight 463.75
MDL Number MFCD01863548
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC359910010

  SDS
thermo fisher scientific
AC359910010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 463.75
Quantity 1g
Packaging Glass bottle
Chemical Name or Material Bis(isopropylcyclopentadienyl)hafnium dichloride
Molecular Formula C16H22Cl2Hf
MDL Number MFCD01863548
CAS 66349-80-2
Health Hazard 1 MOISTURE SENSITIVE
Safety and Handling

MOISTURE SENSITIVE