Please login to your online account to display your discounted pricing

Bis(isopropylcyclopentadienyl)hafnium dichloride, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC359910010

 View more versions of this product

Catalog No. AC359910010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 463.75
Quantity 1g
Packaging Glass bottle
Chemical Name or Material Bis(isopropylcyclopentadienyl)hafnium dichloride
Health Hazard 1 MOISTURE SENSITIVE
CAS 66349-80-2
MDL Number MFCD01863548
Molecular Formula C16H22Cl2Hf

Specifications

Chemical Name or Material Bis(isopropylcyclopentadienyl)hafnium dichloride
CAS 66349-80-2
Molecular Formula C16H22Cl2Hf
Formula Weight 463.75
MDL Number MFCD01863548
Accessories

MOISTURE SENSITIVE

SDS & Safety Documents