Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(hydroxymethyl)phenylphosphine

Chemical Identifiers

Chemical Name or Material Bis(hydroxymethyl)phenylphosphine
CAS 3127-08-0
Molecular Formula C6H5P(CH2OH)2
Formula Weight 170.15
MDL Number MFCD01311823
UN Number UN2811
Synonym (Phenylphosphinidene)dimethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27733MC

 
alfa aesar™
H27733MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH27733ME

 
alfa aesar™
H27733ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.15
Chemical Name or Material Bis(hydroxymethyl)phenylphosphine
Molecular Formula C6H5P(CH2OH)2
MDL Number MFCD01311823
CAS 3127-08-0
UN Number UN2811
Synonym (Phenylphosphinidene)dimethanol
Melting Point 56° to 58°C
Health Hazard 1 Danger
Quantity 100mg
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.