missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(hexylene Glycolato)diboron, TCI America™

$128.34 - $395.70

Chemical Identifiers

CAS 230299-21-5
Molecular Formula C12H24B2O4
Molecular Weight (g/mol) 253.94
MDL Number MFCD03490499
InChI Key UEBSWKNVDRJVHN-UHFFFAOYSA-N
Synonym bis hexylene glycolato diboron, bis hexyleneglycolato diboron, bis hexyleneglycolato diboron, 4,4,6-trimethyl-2-4,4,6-trimethyl-1,3,2-dioxaborinan-2-yl-1,3,2-dioxaborinane, 4,4,4',4',6,6'-hexamethyl-2,2'-bi 1,3,2-dioxaborinane, 2,2'-bi-1,3,2-dioxaborinane, 4,4,4',4',6,6'-hexamethyl, 4,4,4',4',6,6'-hexamethyl-2,2'-bi-1,3,2-dioxaborinane, 4,4,4',4',6,6'-hexamethyl-2,2'-bi 1,3,2-dioxaborinane, pubchem7774, bis hexylene glycolato diboron
PubChem CID 2734616
IUPAC Name 4,4,6-trimethyl-2-(4,4,6-trimethyl-1,3,2-dioxaborinan-2-yl)-1,3,2-dioxaborinane
SMILES B1(OC(CC(O1)(C)C)C)B2OC(CC(O2)(C)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B29061G View Documents TCI America
B29061G
1g Each for $128.34
 
B29065G View Documents TCI America
B29065G
5g Each for $395.70
 
Specifications

Chemical Identifiers

230299-21-5
253.94
UEBSWKNVDRJVHN-UHFFFAOYSA-N
2734616
B1(OC(CC(O1)(C)C)C)B2OC(CC(O2)(C)C)C
C12H24B2O4
MFCD03490499
bis hexylene glycolato diboron, bis hexyleneglycolato diboron, bis hexyleneglycolato diboron, 4,4,6-trimethyl-2-4,4,6-trimethyl-1,3,2-dioxaborinan-2-yl-1,3,2-dioxaborinane, 4,4,4',4',6,6'-hexamethyl-2,2'-bi 1,3,2-dioxaborinane, 2,2'-bi-1,3,2-dioxaborinane, 4,4,4',4',6,6'-hexamethyl, 4,4,4',4',6,6'-hexamethyl-2,2'-bi-1,3,2-dioxaborinane, 4,4,4',4',6,6'-hexamethyl-2,2'-bi 1,3,2-dioxaborinane, pubchem7774, bis hexylene glycolato diboron
4,4,6-trimethyl-2-(4,4,6-trimethyl-1,3,2-dioxaborinan-2-yl)-1,3,2-dioxaborinane

Specifications

White-Yellow
1g
C12H24B2O4
bis hexylene glycolato diboron, bis hexyleneglycolato diboron, bis hexyleneglycolato diboron, 4,4,6-trimethyl-2-4,4,6-trimethyl-1,3,2-dioxaborinan-2-yl-1,3,2-dioxaborinane, 4,4,4',4',6,6'-hexamethyl-2,2'-bi 1,3,2-dioxaborinane, 2,2'-bi-1,3,2-dioxaborinane, 4,4,4',4',6,6'-hexamethyl, 4,4,4',4',6,6'-hexamethyl-2,2'-bi-1,3,2-dioxaborinane, 4,4,4',4',6,6'-hexamethyl-2,2'-bi 1,3,2-dioxaborinane, pubchem7774, bis hexylene glycolato diboron
B1(OC(CC(O1)(C)C)C)B2OC(CC(O2)(C)C)C
253.94
253.94
≥98.0% (GC)
105°C
230299-21-5
MFCD03490499
UEBSWKNVDRJVHN-UHFFFAOYSA-N
4,4,6-trimethyl-2-(4,4,6-trimethyl-1,3,2-dioxaborinan-2-yl)-1,3,2-dioxaborinane
2734616
Crystalline Powder at 20°C
Bis(hexylene Glycolato)diboron
Safety and Handling

Safety and Handling

TSCA : No