Learn More
Bis(heptafluoroisopropyl)ketone, 97%, Thermo Scientific Chemicals
$349.41 - $349.41
Chemical Identifiers
CAS | 813-44-5 |
---|---|
Molecular Formula | C7F14O |
Molecular Weight (g/mol) | 366.054 |
MDL Number | MFCD00042087 |
InChI Key | GRVMOMUDALILLH-UHFFFAOYSA-N |
Synonym | bis perfluoroisopropyl ketone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl pentan-3-one, bis perfluorisopropyl ketone, bis heptafluoroisopropyl ketone, 3-pentanone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl, perfluoroisopropyl ketone, bis heptafluoroisopropyl methanone, cf3 2cfc o cf cf3 2, 3-hexanone, 1,1,1,2,4,4,5,5,6,6,6-undecafluoro-2-trifluoromethyl, tert-butyl 2-oxo-1-pyrrolidinecarboxylat |
PubChem CID | 69941 |
IUPAC Name | 1,1,1,2,4,5,5,5-octafluoro-2,4-bis(trifluoromethyl)pentan-3-one |
SMILES | C(=O)(C(C(F)(F)F)(C(F)(F)F)F)C(C(F)(F)F)(C(F)(F)F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1670806
|
Thermo Scientific Chemicals
L1670806 |
5 g |
Each for $349.41
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
813-44-5 | |
366.054 | |
GRVMOMUDALILLH-UHFFFAOYSA-N | |
69941 | |
C(=O)(C(C(F)(F)F)(C(F)(F)F)F)C(C(F)(F)F)(C(F)(F)F)F |
C7F14O | |
MFCD00042087 | |
bis perfluoroisopropyl ketone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl pentan-3-one, bis perfluorisopropyl ketone, bis heptafluoroisopropyl ketone, 3-pentanone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl, perfluoroisopropyl ketone, bis heptafluoroisopropyl methanone, cf3 2cfc o cf cf3 2, 3-hexanone, 1,1,1,2,4,4,5,5,6,6,6-undecafluoro-2-trifluoromethyl, tert-butyl 2-oxo-1-pyrrolidinecarboxylat | |
1,1,1,2,4,5,5,5-octafluoro-2,4-bis(trifluoromethyl)pentan-3-one |
Specifications
813-44-5 | |
72°C to 73°C | |
MFCD00042087 | |
UN2810 | |
bis perfluoroisopropyl ketone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl pentan-3-one, bis perfluorisopropyl ketone, bis heptafluoroisopropyl ketone, 3-pentanone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl, perfluoroisopropyl ketone, bis heptafluoroisopropyl methanone, cf3 2cfc o cf cf3 2, 3-hexanone, 1,1,1,2,4,4,5,5,6,6,6-undecafluoro-2-trifluoromethyl, tert-butyl 2-oxo-1-pyrrolidinecarboxylat | |
C(=O)(C(C(F)(F)F)(C(F)(F)F)F)C(C(F)(F)F)(C(F)(F)F)F | |
366.054 | |
366.06 | |
Bis(heptafluoroisopropyl)ketone |
1.661 | |
C7F14O | |
5 g | |
1916532 | |
GRVMOMUDALILLH-UHFFFAOYSA-N | |
1,1,1,2,4,5,5,5-octafluoro-2,4-bis(trifluoromethyl)pentan-3-one | |
69941 | |
97% |
Safety and Handling
GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 212-384-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only