missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(ethylenedithio)tetrathiafulvalene, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03955703
Description
It is used as a component for organic superconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Bis(ethylenedithiolo)tetrathiafulvalene | |
238°C (decomposition) | |
MFCD00059710 | |
bis ethylenedithio tetrathiafulvalene, bedt-ttf, 2-5h,6h-1,3 dithiolo 4,5-b 1,4 dithiin-2-ylidene-5h,6h-1,3 dithiolo 4,5-b 1,4 dithiine, bis ethylenedithio tetrathiofulvalene, bis ethylenedithiolo tetrathiafulvalene, bis-ethylenedithio tetrathiafulvalene, bis ethylenedithiolo tetrathiafulvalene 250mg, bi 5,6-dihydro-1,3-dithiolo 4,5-b 1,4-dithiine-2-ylidene, 2-5,6-dihydro-1,3 dithiolo 4,5-b 1,4 dithiin-2-ylidene-5,6-dihydro-1,3 dithiolo 4,5-b 1,4 dithiine | |
LZJCVNLYDXCIBG-UHFFFAOYSA-N | |
2-(5,6-dihydro-[1,3]dithiolo[4,5-b][1,4]dithiin-2-ylidene)-5,6-dihydro-[1,3]dithiolo[4,5-b][1,4]dithiine | |
633432 | |
Powder |
66946-48-3 | |
C10H8S8 | |
1 g | |
Soluble in organic solvents. | |
C1CSC2=C(SC(S2)=C2SC3=C(S2)SCCS3)S1 | |
384.65 | |
384.68 |
Chemical Identifiers
66946-48-3 | |
384.65 | |
LZJCVNLYDXCIBG-UHFFFAOYSA-N | |
633432 | |
C1CSC2=C(SC(S2)=C2SC3=C(S2)SCCS3)S1 |
C10H8S8 | |
MFCD00059710 | |
bis ethylenedithio tetrathiafulvalene, bedt-ttf, 2-5h,6h-1,3 dithiolo 4,5-b 1,4 dithiin-2-ylidene-5h,6h-1,3 dithiolo 4,5-b 1,4 dithiine, bis ethylenedithio tetrathiofulvalene, bis ethylenedithiolo tetrathiafulvalene, bis-ethylenedithio tetrathiafulvalene, bis ethylenedithiolo tetrathiafulvalene 250mg, bi 5,6-dihydro-1,3-dithiolo 4,5-b 1,4-dithiine-2-ylidene, 2-5,6-dihydro-1,3 dithiolo 4,5-b 1,4 dithiin-2-ylidene-5,6-dihydro-1,3 dithiolo 4,5-b 1,4 dithiine | |
2-(5,6-dihydro-[1,3]dithiolo[4,5-b][1,4]dithiin-2-ylidene)-5,6-dihydro-[1,3]dithiolo[4,5-b][1,4]dithiine |
Safety and Handling
TSCA : No
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only