Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(ethylene)(2,4-pentanedionato)rhodium(I), Rh 39.9% min

Chemical Identifiers

Chemical Name or Material Bis(ethylene)(2,4-pentanedionato)rhodium(I)
CAS 12082-47-2
Formula Weight 258.13
MDL Number MFCD00015354
Synonym (Acetylacetonato)bis(ethylene)rhodium(I)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3928803

  SDS
alfa aesar™
3928803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39288MD

  SDS
alfa aesar™
39288MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 258.13
Quantity 1g
MDL Number MFCD00015354
Physical Form Crystalline
Concentration Rh 39.9% min.
CAS 12082-47-2
Solubility Information Soluble in dichloromethane, chloroform
Chemical Name or Material Bis(ethylene)(2,4-pentanedionato)rhodium(I)
Sensitivity Air Sensitive
Synonym (Acetylacetonato)bis(ethylene)rhodium(I)