Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(ethylcyclopentadienyl)magnesium, 98+%

Chemical Identifiers

Chemical Name or Material Bis(ethylcyclopentadienyl)magnesium
CAS 114460-02-5
Formula Weight 210.6
MDL Number MFCD00151375
UN Number UN3392
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3859703

  SDS
alfa aesar™
3859703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3859706

  SDS
alfa aesar™
3859706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38597MD

  SDS
alfa aesar™
38597MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.6
Packaging Ampule
Health Hazard 3 GHS P Statement
P210-P222-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
UN Number UN3392
Health Hazard 2 GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 114460-02-5
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC
MDL Number MFCD00151375
Sensitivity Air & Moisture Sensitive
Density 1.91g/mL
Health Hazard 1 Danger
Chemical Name or Material Bis(ethylcyclopentadienyl)magnesium
Quantity 1g
Assay ≥98%
Physical Form Liquid
Safety and Handling

Danger

GHS P Statement
P210-P222-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.

GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.