missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(diphenylphosphino)methane, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC293610250

 View more versions of this product

Catalog No. AC293610250



Description

Description

Specifications

Specifications

Bis(diphenylphosphino)methane
Glass bottle
118.0°C to 122.0°C
98%
97.5
98%
(C6H5)2PCH2P(C6H5)2
15,1254
XGCDBGRZEKYHNV-UHFFFAOYSA-N
diphenylphosphanylmethyl(diphenyl)phosphane
74952
Fine Crystalline Powder
Authentic
White
25g
2071-20-7
100.0
C25H22P2
MFCD00003537
1,1-bis diphenylphosphino methane, bis diphenylphosphino methane, diphenylphosphanyl methyl diphenylphosphane, dppm, methanediylbis diphenylphosphine, methylenebis diphenylphosphine, phosphine, methylenebis diphenyl, tetraphenylmethylenediphosphine, unii-i5o2hzr38v
C1=CC=C(C=C1)P(CP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
384.39
384.39
98%

Chemical Identifiers

2071-20-7
384.39
XGCDBGRZEKYHNV-UHFFFAOYSA-N
74952
C1=CC=C(C=C1)P(CP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
C25H22P2
MFCD00003537
1,1-bis diphenylphosphino methane, bis diphenylphosphino methane, diphenylphosphanyl methyl diphenylphosphane, dppm, methanediylbis diphenylphosphine, methylenebis diphenylphosphine, phosphine, methylenebis diphenyl, tetraphenylmethylenediphosphine, unii-i5o2hzr38v
diphenylphosphanylmethyl(diphenyl)phosphane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-194-2

SDS Documents