missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Bis(diphenylphosphino)acetylene, 98%, ACROS Organics™
Chemical Identifiers
CAS
5112-95-8
Molecular Formula
C26H20P2
Molecular Weight (g/mol)
394.394
MDL Number
MFCD00003045
InChI Key
FOWZHJNBFVLJGP-UHFFFAOYSA-N
Synonym
1,2-bis diphenylphosphino ethyne, 2-diphenylphosphanyl ethynyl diphenylphosphane, 2-diphenylphosphanylethynyl diphenyl phosphane, acmc-20almm, bis diphenylphosphino acetylene, ethyne-1,2-diylbis diphenylphosphane, ethynylenebis diphenylphosphine, phosphine, 1,2-ethynediylbis diphenyl, phosphine, ethynylenebis diphenyl, pubchem6917
PubChem CID
78789
IUPAC Name
2-diphenylphosphanylethynyl(diphenyl)phosphane
SMILES
C1=CC=C(C=C1)P(C#CP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
View More Specs
Products
1
Catalog Number
Mfr. No.
Quantity
Packaging
Price
Quantity
AC347790050
SDS
View Documents
Thermo Fisher Scientific
347790050
5g
Glass Bottle
N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Specifications
Chemical Identifiers
CAS
5112-95-8
Molecular Weight (g/mol)
394.394
InChI Key
FOWZHJNBFVLJGP-UHFFFAOYSA-N
PubChem CID
78789
SMILES
C1=CC=C(C=C1)P(C#CP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
Molecular Formula
C26H20P2
MDL Number
MFCD00003045
Synonym
1,2-bis diphenylphosphino ethyne, 2-diphenylphosphanyl ethynyl diphenylphosphane, 2-diphenylphosphanylethynyl diphenyl phosphane, acmc-20almm, bis diphenylphosphino acetylene, ethyne-1,2-diylbis diphenylphosphane, ethynylenebis diphenylphosphine, phosphine, 1,2-ethynediylbis diphenyl, phosphine, ethynylenebis diphenyl, pubchem6917
IUPAC Name
2-diphenylphosphanylethynyl(diphenyl)phosphane
Specifications
Infrared Spectrum
Authentic
Color
White
Quantity
5g
CAS Min %
97.5
Assay Percent Range
98%
Linear Formula
(C6 H5 )2 PC≡CP(C6 H5 )2
Synonym
1,2-bis diphenylphosphino ethyne, 2-diphenylphosphanyl ethynyl diphenylphosphane, 2-diphenylphosphanylethynyl diphenyl phosphane, acmc-20almm, bis diphenylphosphino acetylene, ethyne-1,2-diylbis diphenylphosphane, ethynylenebis diphenylphosphine, phosphine, 1,2-ethynediylbis diphenyl, phosphine, ethynylenebis diphenyl, pubchem6917
SMILES
C1=CC=C(C=C1)P(C#CP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
Molecular Weight (g/mol)
394.394
Formula Weight
394.39
Percent Purity
98%
Packaging
Glass Bottle
Melting Point
84.0°C to 88.0°C
CAS
5112-95-8
CAS Max %
100.0
Molecular Formula
C26H20P2
MDL Number
MFCD00003045
InChI Key
FOWZHJNBFVLJGP-UHFFFAOYSA-N
IUPAC Name
2-diphenylphosphanylethynyl(diphenyl)phosphane
PubChem CID
78789
Physical Form
Crystals or Crystalline Powder
Chemical Name or Material
Bis(diphenylphosphino)acetylene, 98%
Safety and Handling
Safety and Handling
GHS H Statement: May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation.
GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 225-842-8
SDS