Please login to your online account to display your discounted pricing

Bis(dimethylthiocarbamyl) Sulfide 97%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  402432500

 View more versions of this product

Catalog No. AC402432500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 208.37
Quantity 250g
Color Yellow
TSCA TSCA
RTECS Number WQ1750000
EINECS Number 202-605-7
Melting Point 106° to 110°C
Particle Size Residue on 500 Micron sieve: <0.001%, residue on 63 micron sieve: <0.05%
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Additional Information oil content: 1.0 to 2.0%
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Name Note 97%
Assay Percent Range 97%
Chemical Name or Material Bis(dimethylthiocarbamyl) sulfide
Linear Formula [(CH3)2NCS)2S
MDL Number MFCD00014870
Molecular Formula C6H12N2S3
CAS 97-74-5
Loss on Drying 0.3% max. (60°C)
Infrared Spectrum Authentic
Synonym Tetramethylthiuram monosulfide
Ash 0.5% max. (900°C)
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Bis(dimethylthiocarbamyl) sulfide
Name Note 97%
CAS 97-74-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H12N2S3
Linear Formula [(CH3)2NCS)2S
Formula Weight 208.37
MDL Number MFCD00014870
Synonym Tetramethylthiuram monosulfide
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.

SDS & Safety Documents