Please login to your online account to display your discounted pricing

Bis(dimethylthiocarbamyl) Sulfide 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Bis(dimethylthiocarbamyl) sulfide
Name Note 97%
CAS 97-74-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H12N2S3
Linear Formula [(CH3)2NCS)2S
Formula Weight 208.37
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC402432500

  SDS
thermo fisher scientific
402432500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 208.37
Quantity 250g
Color Yellow
EINECS Number 202-605-7
TSCA TSCA
Particle Size Residue on 500 Micron sieve: <0.001%, residue on 63 micron sieve: <0.05%
Melting Point 106° to 110°C
RTECS Number WQ1750000
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Additional Information oil content: 1.0 to 2.0%
Chemical Name or Material Bis(dimethylthiocarbamyl) sulfide
Infrared Spectrum Authentic
Loss on Drying 0.3% max. (60°C)
Molecular Formula C6H12N2S3
Linear Formula [(CH3)2NCS)2S
MDL Number MFCD00014870
CAS 97-74-5
Synonym Tetramethylthiuram monosulfide
Health Hazard 1 GHS Signal Word: Warning
Ash 0.5% max. (900°C)
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.