Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(dimethylamino)phosphorochloridate, 94%

Chemical Identifiers

Chemical Name or Material Bis(dimethylamino)phosphorochloridate
CAS 1605-65-8
Linear Formula [(CH3)2N]2P(O)Cl
Formula Weight 170.58
MDL Number MFCD00008302
UN Number UN3265
Beilstein 471430
Synonym Bis(dimethylamino)phosphoryl chloride; N,N,N'N'-Tetramethylphosphorodiamidic chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3927404

  SDS
alfa aesar™
3927404
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3927409

  SDS
alfa aesar™
3927409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.58
MDL Number MFCD00008302
Synonym Bis(dimethylamino)phosphoryl chloride; N,N,N'N'-Tetramethylphosphorodiamidic chloride
Density 1.152g/mL
Odor Pungent
Health Hazard 1 Danger
Linear Formula [(CH3)2N]2P(O)Cl
CAS 1605-65-8
Physical Form Liquid
Flash Point 119° (246°F)
Boiling Point 246° to 247°C
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Refractive Index 1.4679
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Sensitivity Moisture Sensitive
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
UN Number UN3265
Assay 94%
Beilstein 471430
Color Colorless
Quantity 2g
Storage Note 1
  • Packaged under argon
Chemical Name or Material Bis(dimethylamino)phosphorochloridate

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Packaged under argon