Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(diethylamino)chlorophosphine, 94%

Chemical Identifiers

Chemical Name or Material Bis(diethylamino)chlorophosphine
CAS 685-83-6
Molecular Formula C8H20ClN2P
Linear Formula [(C2H5)2N]2PCl
Formula Weight 210.69
MDL Number MFCD00015493
UN Number UN3265
Synonym Bis(diethylamino)phosphorochloridite; Chlorobis(diethylamino)phosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2101906

  SDS
alfa aesar™
B2101906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2101914

  SDS
alfa aesar™
B2101914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2101922

  SDS
alfa aesar™
B2101922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.69
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Synonym Bis(diethylamino)phosphorochloridite; Chlorobis(diethylamino)phosphine
Odor Acrid
Flash Point 101°C (214°F)
MDL Number MFCD00015493
Boiling Point 55° to 57°C (0.2mmHg)
Linear Formula [(C2H5)2N]2PCl
Chemical Name or Material Bis(diethylamino)chlorophosphine
CAS 685-83-6
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Assay 94%
Health Hazard 1 Danger
Quantity 5g
UN Number UN3265
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310-P402
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Molecular Formula C8H20ClN2P
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310-P402
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.