Please login to your online account to display your discounted pricing

Bis(dichlorophosphino)methane 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Bis(dichlorophosphino)methane
CAS 28240-68-8
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula CH2Cl4P2
Linear Formula (Cl2P)2CH2
Formula Weight 217.79
MDL Number MFCD01073767
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383000010

  SDS
thermo fisher scientific
AC383000010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC383000050

  SDS
thermo fisher scientific
AC383000050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 217.79
Physical Form Liquid
Quantity 1g
Color Clear colorless to pale yellow
Density 1.601g/mL
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula CH2Cl4P2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Packaging Glass bottle
NMR Complies, 95% min.
Refractive Index 1.5920 to 1.5950 (20°C, 589nm)
Linear Formula (Cl2P)2CH2
Chemical Name or Material Bis(dichlorophosphino)methane
CAS 28240-68-8
MDL Number MFCD01073767
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.