Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(dicarbonylcyclopentadienyliron), 99%

Chemical Identifiers

Chemical Name or Material Bis(dicarbonylcyclopentadienyliron)
CAS 12154-95-9
Formula Weight 353.93
MDL Number MFCD00135625
UN Number UN3466
Synonym Dicarbonylcyclopentadienyliron dimer; Dicyclopentadienyl-di-iron tetracarbonyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3111406

  SDS
alfa aesar™
3111406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3111414

  SDS
alfa aesar™
3111414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3111422

  SDS
alfa aesar™
3111422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 353.93
Chemical Name or Material Bis(dicarbonylcyclopentadienyliron)
Physical Form Crystalline
Assay 99%
MDL Number MFCD00135625
CAS 12154-95-9
UN Number UN3466
Synonym Dicarbonylcyclopentadienyliron dimer; Dicyclopentadienyl-di-iron tetracarbonyl
Color Red
Melting Point 194°C (dec.)
Health Hazard 1 Danger
Quantity 5g
Sensitivity Air & Moisture Sensitive
Health Hazard 2 GHS H Statement
H228-H300-H310-H330
Flammable solid.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Health Hazard 3 GHS P Statement
P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Packaged under argon
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H300-H310-H330
Flammable solid.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

  • Packaged under argon