missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(dibenzylideneacetone)palladium, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 291970050
Specifications
Bis(dibenzylideneacetone)palladium | |
Black to Brown-Red | |
5g | |
C34H28O2Pd | |
MFCD00051942 | |
1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, palladium 0 bis dibenzylideneacetone, pd dba 2, pubchem14428, tris dibenzylideneacetone dipalladium o | |
UKSZBOKPHAQOMP-SVLSSHOZSA-N | |
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium | |
6505921 | |
Fine Powder |
Glass bottle | |
150.0°C | |
32005-36-0 | |
Pd(C6H5CH=CHC(O)CH=CHC6H5)2 | |
15,302 | |
Solubility in water: insoluble. | |
C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.[Pd] | |
575.02 | |
575.02 | |
16 to 21% (Pd) |
Chemical Identifiers
32005-36-0 | |
575.02 | |
UKSZBOKPHAQOMP-SVLSSHOZSA-N | |
6505921 | |
C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.[Pd] |
C34H28O2Pd | |
MFCD00051942 | |
1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, palladium 0 bis dibenzylideneacetone, pd dba 2, pubchem14428, tris dibenzylideneacetone dipalladium o | |
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
GHS Signal Word: Warning