missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0), TCI America™

$244.45 - $867.80

Chemical Identifiers

CAS 1233717-68-4
Molecular Formula C32H56N2P2Pd
Molecular Weight (g/mol) 637.182
MDL Number MFCD15071402
InChI Key SSPOQURGNAWORH-UHFFFAOYSA-N
Synonym bis di-tert-butyl 4-dimethylaminophenyl phosphine palladium 0, bis 4-n,n-dimethylamino phenyl di-t-butylphosphino palladium 0, bis 4-di-tert-butylphosphanyl-n,n-dimethylaniline palladium
PubChem CID 46900632
IUPAC Name 4-ditert-butylphosphanyl-N,N-dimethylaniline;palladium
SMILES CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.[Pd]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B4837100MG View Documents TCI America
B4837100MG
100mg Each for $244.45
 
B4837500MG View Documents TCI America
B4837500MG
500mg Each for $867.80
 
Specifications

Chemical Identifiers

1233717-68-4
637.182
SSPOQURGNAWORH-UHFFFAOYSA-N
46900632
CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.[Pd]
C32H56N2P2Pd
MFCD15071402
bis di-tert-butyl 4-dimethylaminophenyl phosphine palladium 0, bis 4-n,n-dimethylamino phenyl di-t-butylphosphino palladium 0, bis 4-di-tert-butylphosphanyl-n,n-dimethylaniline palladium
4-ditert-butylphosphanyl-N,N-dimethylaniline;palladium

Specifications

Undesignated
1233717-68-4
MFCD15071402
SSPOQURGNAWORH-UHFFFAOYSA-N
4-ditert-butylphosphanyl-N,N-dimethylaniline;palladium
46900632
Crystal-Powder at 20°C
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0)
100mg
C32H56N2P2Pd
bis di-tert-butyl 4-dimethylaminophenyl phosphine palladium 0, bis 4-n,n-dimethylamino phenyl di-t-butylphosphino palladium 0, bis 4-di-tert-butylphosphanyl-n,n-dimethylaniline palladium
CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.[Pd]
637.182
637.18
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No