missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0), Pd 16.7%

Manufacturer:  Alfa Aesar™ 4600003

 View more versions of this product

Catalog No. AA4600003



Specifications

Specifications

Bis[di-tert-butyl (4-dimethylaminophenyl)phosphine]palladium (0)
C32H56N2P2Pd
SSPOQURGNAWORH-UHFFFAOYSA-N
CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.[Pd]
Pd 16.7%
46900632
637.18
Air Sensitive
1233717-68-4
MFCD15071402
White
4-ditert-butylphosphanyl-N,N-dimethylaniline;palladium
637.182
1g
Crystalline

Chemical Identifiers

1233717-68-4
637.182
SSPOQURGNAWORH-UHFFFAOYSA-N
4-ditert-butylphosphanyl-N,N-dimethylaniline;palladium
C32H56N2P2Pd
MFCD15071402
46900632
CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.[Pd]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents