Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)zirconium hydridochloride, 95%

$144.80 - $519.84

Chemical Identifiers

Chemical Name or Material Bis(cyclopentadienyl)zirconium hydridochloride
CAS 37342-97-5
Molecular Formula C10H11ClZr
Formula Weight 257.87
MDL Number MFCD02089401
UN Number UN3131
Merck Index 14,8396
Synonym Schwartz's Reagent; Zirconocene chloride hydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1516406

  SDS
alfa aesar™
L1516406*
5g Each for $144.80

AAL1516414

  SDS
alfa aesar™
L1516414*
25g Each for $519.84

AAL1516403

  SDS
alfa aesar™
L1516403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 257.87
Chemical Name or Material Bis(cyclopentadienyl)zirconium hydridochloride
Assay 95%
Molecular Formula C10H11ClZr
MDL Number MFCD02089401
CAS 37342-97-5
UN Number UN3131
Melting Point >300°C
Merck Index 14,8396
Synonym Schwartz's Reagent; Zirconocene chloride hydride
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228-H261-H314-H318
Flammable solid.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.

Applications
Bis(cyclopentadienyl)zirconium hydridochloride is used in the conversion of amides to aldehydes. It is also used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas. It is sensitive to moisture, light and air. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H261-H314-H318
Flammable solid.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C