Please login to your online account to display your discounted pricing

Bis(cyclopentadienyl)zirconium dihydride, ACROS Organics™

$220.33 - $220.33

Chemical Identifiers

Chemical Name or Material Bis(cyclopentadienyl)zirconium dihydride
CAS 37342-98-6
Molecular Formula C10H12Zr
Formula Weight 223.43
MDL Number MFCD04126436
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC380140050

  SDS
thermo fisher scientific
380140050
Glass bottle 5g Each for $220.33
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 223.43
Quantity 5g
Color White
Zirconium (Zr) 38.4% min.
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Bis(cyclopentadienyl)zirconium dihydride
MDL Number MFCD04126436
Health Hazard 2 GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Additional Information discolors on exposure to air
Packaging Glass bottle
Molecular Formula C10H12Zr
CAS 37342-98-6

Reduction reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.