missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)zirconium dihydride, 98%

Hydrogenation catalyst for alkenes and alkynes

Manufacturer:  Alfa Aesar™ 4437703

 View more versions of this product

Catalog No. AA4437703



Specifications

Specifications

Bis (cyclopentadienyl)zirconium dihydride
White
37342-98-6
MFCD04126436
bis cyclopenta-2,4-dien-1-yl zirconiumbis ylium dihydride
223.42
98%
Air and moisture sensitive
1g
C10H12Zr
UN3131
It reacts with water.
Powder

Chemical Identifiers

37342-98-6
C10H12Zr
MFCD04126436
bis cyclopenta-2,4-dien-1-yl zirconiumbis ylium dihydride
Safety and Handling

Safety and Handling

GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P223-P231+P232-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H228-H261-H314-H500

DOTInformation : Hazard Class: 4.3; Packaging Group: I

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents