Learn More
Bis(cyclopentadienyl)zirconium chloride hydride, 95%, Thermo Scientific Chemicals
$245.68 - $245.68
Chemical Identifiers
CAS | 37342-97-5 |
---|---|
Molecular Formula | C10H11ClZr |
Molecular Weight (g/mol) | 257.86 |
MDL Number | MFCD02089401 |
InChI Key | VPQMECJLNPUNGN-UHFFFAOYSA-M |
Synonym | zirconocene chloride hydride |
PubChem CID | 131718033 |
IUPAC Name | cyclopenta-1,3-diene;hydride;zirconium(2+);chloride |
SMILES | [H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC207890050
|
Thermo Scientific Chemicals
207890050 |
5 g | Glass bottle |
Each for $245.68
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
37342-97-5 | |
257.86 | |
VPQMECJLNPUNGN-UHFFFAOYSA-M | |
131718033 | |
[H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2] |
C10H11ClZr | |
MFCD02089401 | |
zirconocene chloride hydride | |
cyclopenta-1,3-diene;hydride;zirconium(2+);chloride |
Specifications
37342-97-5 | |
100.0 | |
White to Beige | |
94% min. (Argentometry) | |
C10H11ClZr | |
5 g | |
zirconocene chloride hydride | |
VPQMECJLNPUNGN-UHFFFAOYSA-M | |
cyclopenta-1,3-diene;hydride;zirconium(2+);chloride | |
131718033 | |
95% | |
Bis(cyclopentadienyl)zirconium chloride hydride |
94.0 | |
>300.0°C | |
Authentic | |
Glass bottle | |
MFCD02089401 | |
15, 8533 | |
Solubility in water: reacts with water. | |
[H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2] | |
257.86 | |
257.86 | |
Powder |
Safety and Handling
GHS H Statement
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
GHS Signal Word: Danger
EINECSNumber : 253-479-5
RUO – Research Use Only