Learn More
Bis(cyclopentadienyl)zirconium chloride hydride, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 207890010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Reduction reagentSpecifications
Bis(cyclopentadienyl)zirconium chloride hydride | |
94.0 | |
Authentic | |
Glass bottle | |
MFCD02089401 | |
15, 8533 | |
Solubility in water: reacts with water. | |
[H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2] | |
257.86 | |
257.86 |
37342-97-5 | |
100.0 | |
94% min. (Argentometry) | |
C10H11ClZr | |
1g | |
zirconocene chloride hydride | |
VPQMECJLNPUNGN-UHFFFAOYSA-M | |
cyclopenta-1,3-diene;hydride;zirconium(2+);chloride | |
131718033 | |
95% |
Safety and Handling
GHS H Statement
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
GHS Signal Word: Danger
EINECSNumber : 253-479-5
RUO â Research Use Only