Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)vanadium, sublimed, 95%

Chemical Identifiers

Chemical Name or Material Bis(cyclopentadienyl)vanadium
CAS 1277-47-0
Name Note Sublimed
Formula Weight 181.13
MDL Number MFCD00058735
UN Number UN3181
Synonym Vanadocene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3957203

  SDS
alfa aesar™
3957203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39572MD

  SDS
alfa aesar™
39572MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.13
MDL Number MFCD00058735
Synonym Vanadocene
Health Hazard 3 GHS P Statement
P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Bis(cyclopentadienyl)vanadium
Name Note Sublimed
Physical Form Crystalline
Assay 95%
CAS 1277-47-0
UN Number UN3181
Melting Point 165° to 167°C
Boiling Point 200°C (1mmHg) (subl.)
Quantity 1g
Sensitivity Air & Moisture Sensitive
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H330-H228-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Storage Note 1
  • Packaged under argon
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H228-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Packaged under argon