missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(cyclopentadienyl)vanadium dichloride, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 35341-0050

 View more versions of this product

Catalog No. AC353410050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Bis(cyclopentadienyl)vanadium dichloride
94.0
Authentic
Crystalline Powder
95%
252.04
Dark green
12083-48-6
100.0
Glass bottle
≥94%
C10 H10 Cl2 V
MFCD00014260
5g

Chemical Identifiers

12083-48-6
100.0
C10 H10 Cl2 V
MFCD00014260
≥94%
94.0
95%
252.04
Bis(cyclopentadienyl)vanadium dichloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if inhaled.
Causes serious eye irritation.
Toxic in contact with skin.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 235-150-8

RTECSNumber : YW1580000