missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(cyclopentadienyl)vanadium dichloride, 95%, ACROS Organics™

Chemical Identifiers

CAS 12083-48-6
Molecular Formula C10H10Cl2V
Molecular Weight (g/mol) 252.04
MDL Number MFCD00014260
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC353410050
SDS View Documents
Thermo Fisher Scientific
353410050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

12083-48-6
C10H10Cl2V
252.04
MFCD00014260

Specifications

Authentic
Green
12083-48-6
100.0
C10H10Cl2V
252.04
Crystalline Powder
Bis(cyclopentadienyl)vanadium dichloride
Glass bottle
5g
94.0
94% min. (Argentometry)
MFCD00014260
252.04
95%
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
Causes serious eye irritation.
Toxic in contact with skin.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim

GHS Signal Word: Danger

EINECSNumber : 235-150-8

RTECSNumber : YW1580000