Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)vanadium dichloride, 95%

Chemical Identifiers

Chemical Name or Material Bis(cyclopentadienyl)vanadium dichloride
CAS 12083-48-6
Formula Weight 252.04
MDL Number MFCD00014260
UN Number UN3285
Synonym Vanadocene dichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3958803

 
alfa aesar™
3958803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3958806

 
alfa aesar™
3958806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 252.04
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Bis(cyclopentadienyl)vanadium dichloride
Physical Form Crystalline
Assay 95%
Density 1.60g/mL
MDL Number MFCD00014260
CAS 12083-48-6
UN Number UN3285
Color Green
Melting Point 250°C (dec.)
Synonym Vanadocene dichloride
Quantity 1g
Sensitivity Air & Moisture Sensitive
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Storage Note 1
  • Packaged under argon
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
Odor Pungent
RTECS Number YW1580000

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Packaged under argon