missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)vanadium bromide, 98%

Manufacturer:  Alfa Aesar™ 4436006

 View more versions of this product

Catalog No. AA4436006



Description

Description

  • Ampouled under argon
Specifications

Specifications

Bis (cyclopentadienyl)vanadium bromide
Air and moisture sensitive
221°C to 222°C
64815-29-8
MFCD03701567
QHYHAEAMZXFABY-UHFFFAOYSA-K
cyclopenta-1,3-diene;vanadium;tribromide
132984028
Powder
Ampule
165°C (0.2mmHg)(sublimation)
5g
C10H10Br3V-5
UN3261
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[V].[Br-].[Br-].[Br-]
420.843
261.04
98%

Chemical Identifiers

64815-29-8
420.843
QHYHAEAMZXFABY-UHFFFAOYSA-K
cyclopenta-1,3-diene;vanadium;tribromide
C10H10Br3V-5
MFCD03701567
132984028
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[V].[Br-].[Br-].[Br-]
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

storageNote1 :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

Documents