Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)tungsten dihydride, 98%

Chemical Identifiers

Chemical Name or Material Bis (cyclopentadienyl)tungsten dihydride
CAS 1271-33-6
Molecular Formula C10H12W
Formula Weight 316.06
MDL Number MFCD04126433
UN Number UN3396
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4426603

  SDS
alfa aesar™
4426603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 316.06
Packaging Ampule
Melting Point >350°C
Chemical Name or Material Bis (cyclopentadienyl)tungsten dihydride
Physical Form Crystalline
Health Hazard 2 GHS H Statement
H228-H261-H315-H319-H335
Flammable solid.
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P231+P232-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Color Yellow
MDL Number MFCD04126433
Assay 98%
Quantity 1g
CAS 1271-33-6
UN Number UN3396
Health Hazard 1 Warning
Molecular Formula C10H12W
Sensitivity Air & Moisture Sensitive
DOT Information Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE, FLAMMABLE
Safety and Handling

Warning

GHS P Statement
P210-P231+P232-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS H Statement
H228-H261-H315-H319-H335
Flammable solid.
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.