missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(cyclopentadienyl)titanium(III) chloride, 96%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 96%
Molecular Formula C10H10ClTi
Formula Weight 213.52
MDL Number MFCD07368650
Synonym Nugent's reagent
Chemical Name or Material Bis(cyclopentadienyl)titanium(III) chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC375740050 SDS Thermo Fisher Scientific
AC375740050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Reduction reagent
Description & Specifications

Chemical Identifiers

96%
213.52
Nugent's reagent
C10H10ClTi
MFCD07368650
Bis(cyclopentadienyl)titanium(III) chloride

Specifications

60955-54-6
Authentic
Powder
C10H10ClTi
MFCD07368650
Nugent's reagent
Bis(cyclopentadienyl)titanium(III) chloride
95% min.
Glass bottle
96%
213.52
Green
5g

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger