missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)titanium dichloride

Manufacturer:  Alfa Aesar™ 1254714

 View more versions of this product

Catalog No. AA1254714



Description

Description

  • Packaged under argon
Specifications

Specifications

Bis(cyclopentadienyl)titanium dichloride
Odorless
Red
25g
C10H10Cl2Ti-2
14,9482
MKNXBRLZBFVUPV-UHFFFAOYSA-L
cyclopenta-1,3-diene;titanium(2+);dichloride
124040768
Crystalline
1.60g/mL
Air & Moisture Sensitive
~285°C (decomposition)
1271-19-8
MFCD00003723
Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]
248.957
249

Chemical Identifiers

1271-19-8
248.957
MKNXBRLZBFVUPV-UHFFFAOYSA-L
124040768
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]
C10H10Cl2Ti-2
MFCD00003723
Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
cyclopenta-1,3-diene;titanium(2+);dichloride
Safety and Handling

Safety and Handling

GHS H Statement
H341-H302-H332-H315-H319-H335
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XR2050000

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents