Learn More
Alfa Aesar™ Bis(cyclopentadienyl)ruthenium, Ru 43.2% min
Manufacturer: Alfa Aesar™ 1278103
Description
- Packaged under argon
Specifications
Bis(cyclopentadienyl)ruthenium | |
194°C to 198°C | |
1287-13-4 | |
Ruthenocene | |
231.26 |
Air & Moisture Sensitive | |
1g | |
MFCD00001442 | |
Ru 43.2% min. | |
Crystalline |
Chemical Identifiers
1287-13-4 | |
MFCD00001442 | |
Ruthenocene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
storageNote1 :
- Packaged under argon