missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)nickel, dry

Manufacturer:  Alfa Aesar™ 8789904

 View more versions of this product

Catalog No. AA8789904



Description

Description

  • Packaged under Argon
Specifications

Specifications

Bis (cyclopentadienyl)nickel
Green
2g
1271-28-9
MFCD00001441
bis cyclopentadienyl nickel ii
C1C=CC=[C-]1.C1C=CC=[C-]1.[Ni+2]
188.883
188.9
Air & Moisture Sensitive
171°C to 173°C
Dry
C10H10Ni
UN3391
RQKPFSQDBAZFJV-UHFFFAOYSA-N
cyclopenta-1,3-diene;nickel(2+)
24942185
Crystalline

Chemical Identifiers

1271-28-9
188.883
RQKPFSQDBAZFJV-UHFFFAOYSA-N
24942185
C1C=CC=[C-]1.C1C=CC=[C-]1.[Ni+2]
C10H10Ni
MFCD00001441
bis cyclopentadienyl nickel ii
cyclopenta-1,3-diene;nickel(2+)
Safety and Handling

Safety and Handling

GHS H Statement
H250-H252-H301-H350-H317
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Toxic if swallowed.
May cause cancer.
May cause an allergic skin reaction.

GHS P Statement
P210-P222-P301+P310-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not allow contact with air.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

SDS Documents