Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)magnesium, sublimed, 97%

Chemical Identifiers

Chemical Name or Material Bis (cyclopentadienyl)magnesium
CAS 1284-72-6
Name Note Sublimed
Molecular Formula C10H10Mg
Formula Weight 154.49
MDL Number MFCD00134487
UN Number UN3391
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4343406

  SDS
alfa aesar™
4343406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.49
Packaging Ampule
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC
UN Number UN3391
Name Note Sublimed
Assay 97%
Melting Point 176°C
Boiling Point 290°C
MDL Number MFCD00134487
Color Off-white
Health Hazard 1 Danger
Physical Form Powder
Chemical Name or Material Bis (cyclopentadienyl)magnesium
Quantity 5g
Storage Note 1
  • Ampouled under argon
Health Hazard 2 GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P222-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not allow contact with air.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 1284-72-6
Sensitivity Air & Moisture Sensitive
Molecular Formula C10H10Mg

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P210-P222-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not allow contact with air.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Ampouled under argon