Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)hafnium dichloride

Chemical Identifiers

Chemical Name or Material Bis(cyclopentadienyl)hafnium dichloride
CAS 12116-66-4
Formula Weight 379.59
MDL Number MFCD00001438
UN Number UN3261
Synonym Hafnocene dichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3610503

 
alfa aesar™
3610503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3610506

 
alfa aesar™
3610506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3610514

 
alfa aesar™
3610514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 379.59
Chemical Name or Material Bis(cyclopentadienyl)hafnium dichloride
TSCA TSCA
Physical Form Crystalline
MDL Number MFCD00001438
CAS 12116-66-4
UN Number UN3261
Melting Point 230° to 233°C
Synonym Hafnocene dichloride
Quantity 1g
Sensitivity Air & Moisture Sensitive
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Packaged under argon
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Packaged under argon