Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)dimethylzirconium(IV), 98+%

$125.45 - $395.59

Chemical Identifiers

Chemical Name or Material Bis(cyclopentadienyl)dimethylzirconium(IV)
CAS 12636-72-5
Molecular Formula C12H16Zr
Formula Weight 251.48
MDL Number MFCD01073786
Synonym Dimethylbis(cyclopentadienyl)zirconium(IV); Zirconocene dimethyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2732503

 
alfa aesar™
H2732503
1g Each for $125.45

AAH2732506

 
alfa aesar™
H2732506
5g Each for $395.59
Description & Specifications

Specifications

Formula Weight 251.48
Chemical Name or Material Bis(cyclopentadienyl)dimethylzirconium(IV)
Assay ≥98%
Molecular Formula C12H16Zr
MDL Number MFCD01073786
CAS 12636-72-5
Melting Point ≈170°C
Synonym Dimethylbis(cyclopentadienyl)zirconium(IV); Zirconocene dimethyl
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C