Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)dimethylhafnium(IV), 97+%

$130.03 - $387.24

Chemical Identifiers

Chemical Name or Material Bis(cyclopentadienyl)dimethylhafnium(IV)
CAS 37260-88-1
Molecular Formula C12H16Hf
Formula Weight 338.75
MDL Number MFCD02093015
Synonym Dimethylbis(cyclopentadienyl)hafnium(IV); Hafnocene dimethyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2696203

  SDS
alfa aesar™
H2696203
1g Each for $130.03

AAH2696206

  SDS
alfa aesar™
H2696206
5g Each for $387.24
Description & Specifications

Specifications

Formula Weight 338.75
Chemical Name or Material Bis(cyclopentadienyl)dimethylhafnium(IV)
Assay ≥97%
Molecular Formula C12H16Hf
MDL Number MFCD02093015
CAS 37260-88-1
Melting Point ≈118°C
Synonym Dimethylbis(cyclopentadienyl)hafnium(IV); Hafnocene dimethyl
Quantity 1g