missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(cyclopentadienyl)cobalt

Manufacturer:  Alfa Aesar™ 1685706

 View more versions of this product

Catalog No. AA1685706



Description

Description

  • Packaged under argon
Specifications

Specifications

Bis(cyclopentadienyl)cobalt
176°C to 180°C (decomposition)
1277-43-6
MFCD00013749
Cobaltocene
C1C=CC=[C-]1.C1C=CC=[C-]1.[Co+2]
189.123
189.12
Air & Moisture Sensitive
5g
C10H10Co
UN3181
SNBRJOIYNQIWSZ-UHFFFAOYSA-N
cobalt(2+);cyclopenta-1,3-diene
24942376
Powder

Chemical Identifiers

1277-43-6
189.123
SNBRJOIYNQIWSZ-UHFFFAOYSA-N
24942376
C1C=CC=[C-]1.C1C=CC=[C-]1.[Co+2]
C10H10Co
MFCD00013749
Cobaltocene
cobalt(2+);cyclopenta-1,3-diene
Safety and Handling

Safety and Handling

GHS H Statement
H228-H341-H317
Flammable solid.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
P210-P261-P280-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under argon

SDS Documents