Learn More
Bis(cyclohexanone) oxaldihydrazone, 98%, Thermo Scientific Chemicals
$92.82 - $284.68
Chemical Identifiers
CAS | 370-81-0 |
---|---|
Molecular Formula | C14H22N4O2 |
Molecular Weight (g/mol) | 278.356 |
MDL Number | MFCD00001659 |
InChI Key | DSRJIHMZAQEUJV-UHFFFAOYSA-N |
Synonym | cuprizone, cuprizane, biscyclohexanone oxaldihydrazone, ethanedioic acid, bis cyclohexylidenehydrazide, oxalic acid bis cyclohexylidenehydrazide, biscyclohexanone oxalyldihydrazone, bis cyclohexanone oxaldihydrazone, cuprizon, unii-5n16u7e0ao, oxalic acid, bis cyclohexylidenehydrazide |
PubChem CID | 9723 |
IUPAC Name | N,N'-bis(cyclohexylideneamino)oxamide |
SMILES | C1CCC(=NNC(=O)C(=O)NN=C2CCCCC2)CC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1062814
|
Thermo Scientific Chemicals
A1062814 |
25 g |
Each for $92.82
|
|
|||||
AAA1062822
|
Thermo Scientific Chemicals
A1062822 |
100 g |
Each for $284.68
|
|
|||||
Description
Bis(cyclohexanone) oxaldihydrazone is used in the quantitative spectroscopic determination of copper. It is also essential for the experimental study of mitochondrial metabolism. It is used to study the human demyelinating diseases such as multiple sclerosis and components of neuroinflammation. Further, it acts as ligand and catalyzes Ullmann-type C-N bond forming reactions in the presence of copper salt.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBis(cyclohexanone) oxaldihydrazone is used in the quantitative spectroscopic determination of copper. It is also essential for the experimental study of mitochondrial metabolism. It is used to study the human demyelinating diseases such as multiple sclerosis and components of neuroinflammation. Further, it acts as ligand and catalyzes Ullmann-type C-N bond forming reactions in the presence of copper salt.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
370-81-0 | |
278.356 | |
DSRJIHMZAQEUJV-UHFFFAOYSA-N | |
9723 | |
C1CCC(=NNC(=O)C(=O)NN=C2CCCCC2)CC1 |
C14H22N4O2 | |
MFCD00001659 | |
cuprizone, cuprizane, biscyclohexanone oxaldihydrazone, ethanedioic acid, bis cyclohexylidenehydrazide, oxalic acid bis cyclohexylidenehydrazide, biscyclohexanone oxalyldihydrazone, bis cyclohexanone oxaldihydrazone, cuprizon, unii-5n16u7e0ao, oxalic acid, bis cyclohexylidenehydrazide | |
N,N'-bis(cyclohexylideneamino)oxamide |
Specifications
370-81-0 | |
C14H22N4O2 | |
25 g | |
cuprizone, cuprizane, biscyclohexanone oxaldihydrazone, ethanedioic acid, bis cyclohexylidenehydrazide, oxalic acid bis cyclohexylidenehydrazide, biscyclohexanone oxalyldihydrazone, bis cyclohexanone oxaldihydrazone, cuprizon, unii-5n16u7e0ao, oxalic acid, bis cyclohexylidenehydrazide | |
DSRJIHMZAQEUJV-UHFFFAOYSA-N | |
N,N'-bis(cyclohexylideneamino)oxamide | |
9723 | |
98% |
208°C to 212°C | |
MFCD00001659 | |
2388004 | |
Soluble in alcohol. Insoluble in water. | |
C1CCC(=NNC(=O)C(=O)NN=C2CCCCC2)CC1 | |
278.356 | |
278.36 | |
Bis (cyclohexanone) oxaldihydrazone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 206-729-2
RTECSNumber : RO2520000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only