Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(Boc-aminooxy)acetic acid, 98%

Chemical Identifiers

Chemical Name or Material Bis(Boc-aminooxy)acetic acid
CAS 293302-31-5
Linear Formula [(CH3)3COCONHO]2CHCOOH
Formula Weight 322.32
MDL Number MFCD06410966
Synonym Bis-Boc-Aoa
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6287903

  SDS
alfa aesar™
H6287903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6287906

  SDS
alfa aesar™
H6287906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62879MD

  SDS
alfa aesar™
H62879MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 322.32
Chemical Name or Material Bis(Boc-aminooxy)acetic acid
Physical Form Powder
Assay 98%
Linear Formula [(CH3)3COCONHO]2CHCOOH
MDL Number MFCD06410966
CAS 293302-31-5
Color White
Synonym Bis-Boc-Aoa
Quantity 1g

Applications
Bis(Boc-aminooxy)acetic acid employed as a protecting reagent and intermediate pharmaceuticals and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.